missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LZIC Monoclonal Antibody (OTI2D3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505939S
This item is not returnable.
View return policy
Description
LZIC Monoclonal antibody specifically detects LZIC in Human, Mouse samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| LZIC | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q80UK0, Q86VW0 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 228071, 91404 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Immunocytochemistry, Western Blot | |
| OTI2D3 | |
| Unconjugated | |
| Sestd1 | |
| 1810030J04Rik; AW047580; EGK_00210; leucine zipper and CTNNBIP1 domain containing; leucine zipper and CTNNBIP1 domain-containing protein; Leucine zipper and ICAT homologous domain-containing protein; leucine zipper domain and ICAT homologous domain containing; LZIC; Protein LZIC | |
| Full length human recombinant protein of human SESTD1 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human, Mouse | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction