missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LZTFL1 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97499B
This item is not returnable.
View return policy
Description
LZTFL1 Polyclonal specifically detects LZTFL1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| LZTFL1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Biotin | |
| FLJ36386, leucine zipper transcription factor-like 1, leucine zipper transcription factor-like protein 1 | |
| Produced in rabbits immunized with E. coli-derived Human LZTFL1 fragment. | |
| 0.1 mL | |
| Epigenetics | |
| 54585 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction