missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LZTR1 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
€ 213.00 - € 507.00
Specifications
| Antigen | LZTR1 |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30232477
|
Novus Biologicals
NBP3-33513-100ul |
100 μL |
€ 507.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30228261
|
Novus Biologicals
NBP3-33513-20ul |
20 μL |
€ 213.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LZTR1 Monoclonal antibody specifically detects LZTR1 in Human,Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| LZTR1 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| 8216 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| leucine-zipper-like regulator-1, leucine-zipper-like transcription regulator 1, leucine-zipper-like transcriptional regulator 1, LZTR-1, MGC21205, TCFL2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 300-545 of human LZTR1 (NP_006758.2).,, Sequence:, GGAADNTLPNELHCYDVDFQTWEVVQPSSDSEVGGAEVPERACASEEVPTLTYEERVGFKKSRDVFGLDFGTTSAKQPTQPASELPSGRLFHAAAVISDAMYIFGGTVDNNIRSGEMYRFQFSCYPKCTLHEDYGRLWESRQFCDVEFVLGEKEECVQGHVAIVTARSRWLRRKITQARERLAQKLEQEAAPVPREAPGVAAGGARPPLLHVAIREAEARPFEVLMQFLYTDKIKYPRKGHVEDVL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title