missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LZTS1 Monoclonal Antibody (OTI3D12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810241
This item is not returnable.
View return policy
Description
This gene encodes a tumor suppressor protein that is ubiquitously expressed in normal tissues. In uveal melanomas, expression of this protein is silenced in rapidly metastasizing and metastatic tumor cells but has normal expression in slowly metastasizing or nonmetastasizing tumor cells. This protein may have a role in cell-cycle control by interacting with the Cdk1/cyclinB1 complex. This gene is located on chromosomal region 8p22. Loss of heterozygosity (LOH) in the 8p arm is a common characteristic of many types of cancer.Specifications
| LZTS1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9Y250 | |
| LZTS1 | |
| Full length human recombit protein of human LZTS1 produced in HEK293T Cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI3D12 | |
| Unconjugated | |
| LZTS1 | |
| F37, FEZ1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 11178 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction