missing translation for 'onlineSavingsMsg'
Learn More

M-Benzyl-N-trifluoroacetyl-L-tryptophan Methyl Ester, TRC

Product Code. 30342964
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30342964 1 g 1g
30332320 250 mg 250mg
30337255 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30342964 Supplier Toronto Research Chemicals Supplier No. TRCB3161451G
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material M-Benzyl-N-trifluoroacetyl-L-tryptophan Methyl Ester
Analyte or Component Names M-Benzyl-N-trifluoroacetyl-L-tryptophan Methyl Ester
Shipping Condition Room Temperature
Molecular Formula C21 H19 F3 N2 O3
InChI Formula InChI=1S/C21H19F3N2O3/c1-29-19(27)18(11-15-12-25-17-10-6-5-9-16(15)17)26(20(28)21(22,23)24)13-14-7-3-2-4-8-14/h2-10,12,18,25H,11,13H2,1H3/t18-/m0/s1
Solution Type Neat
SMILES COC(=O)[C@H](Cc1c[nH]c2ccccc12)N(Cc3ccccc3)C(=O)C(F)(F)F
IUPAC Name methyl (2S)-2-[benzyl-(2,2,2-trifluoroacetyl)amino]-3-(1H-indol-3-yl)propanoate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 404.382
Quantity 1 g
Formula Weight 404.135
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.