missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ M-cadherin Polyclonal Antibody
GREENER_CHOICE

Product Code. 17138683
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17138683 100 μL 100µL
17149583 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17138683 Supplier Invitrogen™ Supplier No. PA5110758

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: LSIVKALDYE SCEHYELKVS VQNEAPLQAA ALRAERGQAK VRVHVQDTNE PPVFQENPLR TSLAEGAPPG TLVAT Highest antigen sequence indentity to the following orthologs: Rat - 79%, Mouse - 80%.

Cadherins are a multigene family of Ca++-dependent cell adhesion molecules. They are transmembrane glycoproteins consisting of an extracellular domain, which mediates Ca++-dependent intercellular adhesion by homophilic interactions, a transmembrane region and a cytoplasmic domain. The extracellular domain is divided into a series of subdomains designated EC1-EC5. Homolgies between different members of the cadherin family are most prominent in the cytoplasmic domain and in EC1 and EC2 and much less so in EC5 of the extracellular domain and in the transmembrane region. The binding properties and specificities of the adhesive function are located in the N-terminal part of the molecules. Four members of the cadherin family have been identified and molecularly cloned from mammalian cells. These include the neuronal (N), epithelial (E), placental (P) and muscle (M) cadherins. M-cadherin is not found in fibroblasts but is expressed at low level in myoblasts and is upregulated following induction of myotube formation, suggesting a specific function in skeletal muscle cell differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen M-cadherin
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CDH15
Gene Accession No. P55291
Gene Alias AI323380; cadherin 15; cadherin 15, type 1, M-cadherin (myotubule); cadherin-14; Cadherin-15; cadherin-3; Cdh14; Cdh15; CDH3; CDHM; CRSBP-1; HAR; LYVE-1; M cadherin; Mcad; M-cadherin; MRD3; Muscle cadherin; muscle-cadherin; XLKD1
Gene Symbols CDH15
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human M-cadherin. Recombinant protein control fragment (Product #RP-90195).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1013
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.