missing translation for 'onlineSavingsMsg'
Learn More
Learn More
M-CSF Antibody (26786), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB2161Y
This item is not returnable.
View return policy
Description
M-CSF Monoclonal antibody specifically detects M-CSF in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| M-CSF | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 26786 | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| colony stimulating factor 1 (macrophage), CSF-1, Lanimostim, macrophage colony stimulating factor, macrophage colony-stimulating factor 1, M-CSF, MCSFlanimostim, MGC31930 | |
| E. coli-derived recombinant human M-CSF | |
| 0.5 mL | |
| Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Stem Cell Markers | |
| 1435 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction