Learn More
M-CSFR/CD115 Antibody (12-3A3-1B10), FITC, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals NBP1-43362F
Product Code. 18577370
  • ONLINE EXCLUSIVE PRICE
    € 593.00
    € 560.70 / 0.10mL
    Save € 32.30
    5% Off
Description
M-CSFR/CD115 Monoclonal antibody specifically detects M-CSFR/CD115 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
M-CSFR/CD115
Monoclonal
FITC
PBS
Rat
Protein A or G purified
RUO
Primary
Human
Purified
Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
12-3A3-1B10
Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin
CD115, CD115 antigen, C-FMS, colony stimulating factor 1 receptor, CSF-1-R, CSFR, EC 2.7.10.1, FMS proto-oncogene, FMSFIM2, macrophage colony stimulating factor I receptor, macrophage colony-stimulating factor 1 receptor, McDonough feline sarcoma viral (v-fms) oncogene homolog, Proto-oncogene c-Fms
The immunogen for this antibody was CD115.
0.1 mL
Myeloid derived Suppressor Cell, Phospho Specific, Protein Kinase
1436
Store at 4C in the dark.
IgG2a κ