missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ M-CSFR/CD115 Antibody (LY3022855), Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28653-100ug
This item is not returnable.
View return policy
Description
M-CSFR/CD115 Monoclonal antibody specifically detects M-CSFR/CD115 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| M-CSFR/CD115 | |
| Monoclonal | |
| Unconjugated | |
| P07333 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| LY3022855 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| CD115, CD115 antigen, C-FMS, colony stimulating factor 1 receptor, CSF-1-R, CSFR, EC 2.7.10.1, FMS proto-oncogene, FMSFIM2, macrophage colony stimulating factor I receptor, macrophage colony-stimulating factor 1 receptor, McDonough feline sarcoma viral (v-fms) oncogene homolog, Proto-oncogene c-Fms | |
| CSF1R/ M-CSFR/ CD115 | |
| 100 μg | |
| Myeloid derived Suppressor Cell, Phospho Specific, Protein Kinase | |
| 1436 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction