missing translation for 'onlineSavingsMsg'
Learn More
Learn More
M13 bacteriophage Antibody (E1), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-66601F
This item is not returnable.
View return policy
Description
M13 bacteriophage Monoclonal antibody specifically detects M13 bacteriophage in Virus samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry
Specifications
| M13 bacteriophage | |
| Monoclonal | |
| FITC | |
| Coat protein A; Coat protein B; Coat protein C polypeptide I; Coat protein C polypeptide II; Coat protein D; G3P; III; IX; M13 coat protein A; M13 coat protein B; M13 coat protein C polypeptide I; M13 coat protein C polypeptide II; M13 coat protein D; M13 | |
| Hybridoma produced by the fusion of splenocytes from mice immunized with isolated M13 phage coat proteins and mouse myeloma cells. | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG3 |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| E1 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction