Learn More
Applied Biosystems™ M13 Sequencing Forward Primer, HPLC
Shop All Applied Biosystems Products
Click to view available options
:
1 each
Description
Use our tool to find and order pre-designed primers for PCR and Sanger sequencing. Free of SNPs and dimers, target-specific, and suitable for universal PCR conditions, these primers can be ordered unmodified, M13-tailed, HPLC-purified, or desalted. All primers are checked by mass spectrometry.