missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MA2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17219503 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17219503 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17219503 Supplier Invitrogen™ Supplier No. PA5117001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Samples: 293T, Jurkat, Rat brain Immunogen sequence: MALALLEDWC RIMSVDEQKS LMVTGIPADF EEAEIQEVLQ ETLKSLGRYR LLGKIFRKQE NANAVLLELL EDTDVSAIPS EVQGKGGVWK VIFKTPNQDT EFLERLNLFL EKEGQTVSGM FRALGQEGVS PATVPCISPE.

In patients with cancer, symptoms of limbic and brain-stem dysfunction may result from a para-neoplastic disorder. Two proteins, Ma1 and Ma2, were discovered by presence of autoantibodies against them. Antibodies to Ma1 and Ma2 proteins identify a paraneoplastic disorder that affects the limbic system, brain stem, and cerebellum. These autoantibodies were used to clone Ma2, which codes for a protein (Ma2) that was recognized by serum from the 10 patients, but not by serum from 344 control subjects. Ma2 was selectively expressed by normal brain tissue and by the testicular tumors of the patients. Ma2 shares homology with Ma1, a brain-testis-cancer gene related to other para-neoplastic syndromes and tumors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MA2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.26 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene PNMA2
Gene Accession No. Q9UL42
Gene Alias 40 kDa neuronal protein; A830049P17Rik; KIAA0883; MA2; mKIAA0883; MM2; onconeuronal antigen MA2; Paraneoplastic antigen Ma2; paraneoplastic antigen Ma2 homolog; paraneoplastic Ma antigen 2; paraneoplastic neuronal antigen MM2; Pnma2; retrotransposon gag domain containing 2; RGAG2
Gene Symbols PNMA2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-140 of human PNMA2 (NP_0091881).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10687, 305977
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.