missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAATS1 Antibody (OTI6G1), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72309JF646
This item is not returnable.
View return policy
Description
MAATS1 Monoclonal specifically detects MAATS1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| MAATS1 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MAATS1 | |
| Recombinant protein fragment corresponding to amino acids 1-313 of human C3orf15 (NP_203528) produced in E.coli. | |
| 0.1 mL | |
| 89876.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI6G1 | |
| Western Blot, Immunohistochemistry | |
| AAT1, AAT-1, AAT1alpha, AAT1-alpha, AAT1DKFZp781A2221, AMY-1-associating protein expressed in testis 1, C3orf15, chromosome 3 open reading frame 15, MYCBP/AMY-1-associated, testis expressed 1, MYCBP-binding protein, SPATA26 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction