missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAB21L1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566289
This item is not returnable.
View return policy
Description
Immunogen sequence: SPTEFEVVLYL NQMGVFNFVD DGSLPGCAVL KLSDGRKRSM SLWVEFITAS GYLSARKIRS RFQTLVAQAV DKCSYRDVVK Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
This gene is similar to the MAB-21 cell fate-determining gene found in C. elegans. It may be involved in eye and cerebellum development, and it has been proposed that expansion of a trinucleotide repeat region in the 5' UTR may play a role in a variety of psychiatric disorders.
Specifications
| MAB21L1 | |
| Polyclonal | |
| Unconjugated | |
| MAB21L1 | |
| AW047968; CAGR1; Mab21; mab-21 like 1; MAB21L1; mab-21-like 1 (C. elegans); mab-21-like protein 1; mb211; Nbla00126; protein mab-21-like 1; Putative nucleotidyltransferase MAB21L1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 4081 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q13394 | |
| MAB21L1 | |
| Recombinant Human MAB21L1. Recombinant protein control fragment (Product #RP-105636). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction