Learn More
Hellma Macro Cuvette, 402-OG
Product Code. p-7160240
Click to view available options
Capacity (Metric):
30,000 μL
6000 μL
Path Length:
10 mm
50 mm
Unit Size:
Each
Product Code. 10065991
Brand: Hellma 4021010
Only null left
Add to basket
Add to basket
Optical glass absorption assay cuvette with a PTFE lid.
  • Optical glass construction
  • VIS range
  • PTFE lid included
Specifications
Capacity (Metric) 6000 μL
Material Optical Glass
Closure Material PTFE
Width (Metric) Interior 18.5 mm
Thickness (Metric) Base thickness: 2.5 mm
Includes PTFE Lid
Quantity 1/Pk
Path Length 10 mm
Wavelength Range 360 to 2500 nm
Height (Metric) 40 mm
Width (Metric) 23.6 mm