missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macro H2A.2 Antibody (CL5263), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61156-25ul
This item is not returnable.
View return policy
Description
Macro H2A.2 Monoclonal specifically detects Macro H2A.2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Macro H2A.2 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.2), containing 40% glycerol with 0.02% Sodium Azide | |
| MACROH2A2 | |
| This antibody was developed against a synthetic peptide corresponding to the following amino acid sequence:EKRGRKATSGKKGGK | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| CL5263 | |
| Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| core histone macro-H2A.2, core histone macroH2A2.2, H2A histone family, member Y2, Histone macroH2A2, macroH2A2, mH2A2 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 55506 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction