missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macrophage Antibody (RM0029-11H3), HRP, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-12436H
This item is not returnable.
View return policy
Description
Macrophage Monoclonal specifically detects Macrophage in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Macrophage | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| Rat | |
| Protein G purified | |
| Primary | |
| Store at 4C in the dark. | |
| IgG2a |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| RM0029-11H3 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| EC 5.3.2.1, EC 5.3.3.12, GIFmacrophage migration inhibitory factor, GLIF, Glycosylation-inhibiting factor, L-dopachrome isomerase, L-dopachrome tautomerase, macrophage migration inhibitory factor (glycosylation-inhibiting factor), MMIF, Phenylpyruvate tautomerase | |
| Isolated mouse macrophages | |
| 0.1 mL | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction