missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macrophage and Histiocytoma Marker Antibody (D11), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11429AF532
This item is not returnable.
View return policy
Description
Macrophage and Histiocytoma Marker Monoclonal antibody specifically detects Macrophage and Histiocytoma Marker in Human, Mouse (Negative), Porcine (Negative), Rat (Negative) samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Macrophage and Histiocytoma Marker | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Membrane preparation from human hepatocytes | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| D11 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Human, Mouse (Negative), Porcine (Negative), Rat (Negative) | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction