missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macrophage Marker Antibody (5/9), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50482MFV500
This item is not returnable.
View return policy
Description
Macrophage Marker Monoclonal antibody specifically detects Macrophage Marker in Human samples. It is validated for Immunohistochemistry
Specifications
| Macrophage Marker | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| A cell suspension containing osteoclasts from osteoclastomas. | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG1 |
| Immunohistochemistry | |
| 2024-05-09 00:00:00 | |
| Immunohistochemistry | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction