missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macrophage Mouse anti-Human, Clone: LN-5, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB14659.100ug
This item is not returnable.
View return policy
Description
Mouse monoclonal antibody raised against human macrophage.
- High specificity
- High reactivity
Specifications
| Macrophage | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (0.5-1 ug/million cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| Mouse | |
| PEG precipitation | |
| RUO | |
| Human | |
| Liquid |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| LN-5 | |
| Mouse monoclonal antibody raised against human macrophage. | |
| In PBS (0.05% sodium azide). | |
| Human peripheral blood mononuclear cells. | |
| 100 μg | |
| Primary | |
| Store at 4°C. | |
| IgM κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction