missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macrophage Rat anti-Mouse, APC, Clone: RM0029-11H3, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-12436APC-0.1ML
This item is not returnable.
View return policy
Description
Macrophage Monoclonal specifically detects Macrophage in Mouse samples. It is validated for Flow Cytometry.
Specifications
| Macrophage | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| Rat | |
| >95% pure by SDS-PAGE | |
| RUO | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| RM0029-11H3 | |
| Flow Cytometry | |
| EC 5.3.2.1, EC 5.3.3.12, GIFmacrophage migration inhibitory factor, GLIF, Glycosylation-inhibiting factor, L-dopachrome isomerase, L-dopachrome tautomerase, macrophage migration inhibitory factor (glycosylation-inhibiting factor), MMIF, Phenylpyruvate tautomerase | |
| Isolated mouse macrophages | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction