Learn More
Invitrogen™ Macrophages (Mature) Monoclonal Antibody (25F9), Biotin
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA549062
Description
The antibody is associated with fully differentiated tissue macrophages both in normal and in diseased tissues, independently of the presence or absence of inflammation. The antigen is absent on freshly isolated monocytes and other blood cells. After 6 to 7 days culture human monocytes become positive. Some melanoma and carcinoma cell lines are also positive. Furthermore the monoclonal antibody 25F9 cross reacts with a subpopulation of macrophages of rhesus monkey, pig alveolar macrophages and Kupffer cells.
Macrophages are a type of white blood cell that plays a crucial role in the immune system. They are responsible for detecting, engulfing, and destroying foreign substances, such as bacteria and viruses, as well as dead cells and debris. Mature macrophages are found in various tissues throughout the body and are highly specialized to perform specific functions depending on their location. For example, alveolar macrophages are found in the lungs and are responsible for removing foreign particles that are inhaled, while Kupffer cells are found in the liver and are responsible for removing old or damaged red blood cells. Understanding the function and behavior of macrophages is crucial for developing new treatments for diseases such as cancer, autoimmune disorders, and infections.
Specifications
| Macrophages (Mature) | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 0.1% BSA and 0.02% sodium azide | |
| Macrophages; Mature Macrophages | |
| The monoclonal antibody 25F9 recognizes a protein of 86 kD on the cell surface and within the cytoplasm of mature macrophages. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
| 25F9 | |
| Biotin | |
| P29466, P48729, Q8N752 | |
| Mouse | |
| Protein G | |
| RUO | |
| 122011, 1452, 834 | |
| 4°C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.