missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAD2L1-binding protein Antibody (OTI3C11), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72566PCP
This item is not returnable.
View return policy
Description
MAD2L1-binding protein Monoclonal antibody specifically detects MAD2L1-binding protein in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| MAD2L1-binding protein | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| OTI3C11 | |
| Western Blot, Immunohistochemistry | |
| Caught by MAD2 protein, CMT2MGC11282, dJ261G23.1, KIAA0110RP1-261G23.6, MAD2L1 binding protein, MAD2L1-binding protein | |
| Full length human recombinant protein of human MAD2L1BP (NP_055443) produced in E.coli. | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 9587 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction