missing translation for 'onlineSavingsMsg'
Learn More

MAD2L2 Antibody, Novus Biologicals™

Product Code. 18612579 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18612579 100 μg 100µL
18629996 50 μg 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18612579 Supplier Novus Biologicals Supplier No. NBP312688100ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MAD2L2 Polyclonal antibody specifically detects MAD2L2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAD2L2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation PBS, pH7.0
Gene Alias hREV7, MAD2 mitotic arrest deficient-like 2 (yeast), MAD2BREV7 homolog, MAD2-like protein 2, Mitotic arrest deficient 2-like protein 2, mitotic arrest deficient homolog-like 2, mitotic spindle assembly checkpoint protein MAD2B, REV7yeast, homolog)-like 2
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human MAD2L2 fragment
Purification Method Antigen and protein A Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 10459
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.