Learn More
MAD2L2 Mouse anti-Human, Unconjugated, Clone: 2A4C2, Proteintech
Product Code. 16829535 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16829535 20 μL 20µL
16819535 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16829535 Supplier Proteintech Supplier No. 671001IG20UL
Mouse Monoclonal Antibody
The MAD2L2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MAD2L2. This antibody recognizes human, mouse, rat antigen. The MAD2L2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Antigen MAD2L2
Applications Western Blot, Immunohistochemistry, ELISA
Classification Monoclonal
Clone 2A4C2
Concentration 1000 μg/mL
Conjugate Unconjugated
Host Species Mouse
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 10459
Target Species Human, Rat, Mouse
Isotype IgG2b