missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAD4 Antibody (OTI7F7), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72567AF532
This item is not returnable.
View return policy
Description
MAD4 Monoclonal specifically detects MAD4 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| MAD4 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| MXD4 | |
| Full length recombinant protein of human MAD4 (NP_006445) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| OTI7F7 | |
| Western Blot, Immunohistochemistry | |
| BHLHC12, bHLHc12Max-interacting transcriptional repressor MAD4, Class C basic helix-loop-helix protein 12, Mad4 homolog, MAD4Max-associated protein 4, MAX dimerization protein 4, Max dimerizer 4, MST149, MSTP149, MXD4 MAX dimerization protein 4 | |
| Mouse | |
| Affinity Purified | |
| Cell Cycle and Replication | |
| 10608.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction