missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAdCAM-1 Antibody (683715), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB6056CL1
This item is not returnable.
View return policy
Description
MAdCAM-1 Monoclonal antibody specifically detects MAdCAM-1 in Human samples. It is validated for Flow Cytometry,CyTOF
Specifications
| MAdCAM-1 | |
| Monoclonal | |
| CoraFluor 1 | |
| hMAdCAM-1, MACAM1, MAdCAM-1, mucosal addressin cell adhesion molecule 1, mucosal addressin cell adhesion molecule-1, mucosal vascular addressin cell adhesion molecule 1 | |
| Mouse myeloma cell line NS0-derived recombinant human MAdCAM-1, Leu21-Gln333, Accession # AAY82472 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 683715 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 8174 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction