missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MAdCAM-1 Polyclonal Antibody

Product Code. 16541242
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16541242 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16541242 Supplier Invitrogen™ Supplier No. PA598417

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MAdCAM-1 Polyclonal Antibody for ELISA, ICC/IF, Western Blot

The protein encoded by this gene is an endothelial cell adhesion molecule that interacts preferentially with the leukocyte beta7 integrin LPAM-1 (alpha4beta7), L-selectin, and VLA-4 (alpha4beta1) on myeloid cells to direct leukocytes into mucosal and inflamed tissues. It is a member of the immunoglobulin family and is similar to ICAM1 and VCAM1. At least seven alternatively spliced transcripts encoding different protein isoforms have been found for this gene, but the full-length nature of some variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAdCAM-1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene MADCAM1
Gene Accession No. Q13477
Gene Alias AV211525; hMAdCAM-1; MACAM1; MADCAM1; MAdCAM-1; mMAdCAM-1; mucosal addressin cell adhesion molecule 1; mucosal addressin cell adhesion molecule-1; mucosal vascular addressin cell adhesion molecule 1; rMAdCAM-1
Gene Symbols MADCAM1
Host Species Rabbit
Immunogen Recombinant Human Mucosal addressin cell adhesion molecule 1 protein (19-317AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8174
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.