missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MADCAM1 Monoclonal Antibody (OTI1B7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA507035
This item is not returnable.
View return policy
Description
MAdCAM-1 protein is an endothelial cell adhesion molecule that interacts preferentially with the leukocyte beta7 integrin LPAM-1 (alpha4beta7), L-selectin, and VLA-4 (alpha4beta1) on myeloid cells to direct leukocytes into mucosal and inflamed tissues. It is a member of the immunoglobulin family and is similar to ICAM1 and VCAM1. At least seven alternatively spliced transcripts encoding different protein isoforms have been found for this gene, but the full-length nature of some variants has not been determined.Specifications
| MADCAM1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O70540, Q13477, Q61826 | |
| Madcam1 | |
| Full length human recombit protein of human MADCAM1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Canine, Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI1B7 | |
| Unconjugated | |
| MADCAM1 | |
| MACAM1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 17123, 485103, 54266, 8174 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction