missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MADD Recombinant Rabbit Monoclonal Antibody (JE63-62)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA544786

Product Code. 17848752

  • € 502.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MADD Recombinant Monoclonal Antibody for Western Blot

Tumor necrosis factor receptor 1 (TNFR1) contains a death receptor domain that mediates interactions involved in downstream signaling of TNF. A novel protein, MADD (mitogen activated protein (MAP)-kinase activating death domain protein) has recently been shown to associate with the death domain of TNFR1 through its own C-terminal death domain. This interaction is critical for TNFR1 signal generation. Overexpression of MADD activates the MAP (Mitogen Activated Protein) kinases extracellular signal-related kinase (ERK) and c-Jun N-terminal kinase (JNK) and induces the phosphorylation of cytosolic phospholipase A2. This discovery suggests that MADD links tumor necrosis factor TNF and MAP kinase activation and arachidonic acid release. MADD has a 98% homology with another protein, differentially expressed in neoplasmic vs. normal cells (DENN). DENN is a substrate for JNK3, and is important in the hypoxia and stress induced intracellular signalling pathways. MADD also has a 93% sequence conservation with a GDP/GTP exchange protein, Rab3.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MADD
Recombinant Monoclonal
1 mg/mL
TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Q80U28, Q8WXG6
MADD
Synthetic peptide within human DENN aa 1,598-1,647/1,647.
100 μL
Primary
Human, Mouse
Antibody
IgG
Western Blot
JE63-62
Unconjugated
MADD
9630059K23Rik; DENN; differentially expressed in normal and neoplastic cells; IG20; Insulinoma glucagonoma clone 20; insulinoma-glucagonoma protein 20; Kiaa0358; Madd; MAP kinase activating death domain; MAP kinase-activating death domain protein; MAP-kinase activating death domain; rab3 GDP/GTP exchange factor; Rab3 GDP/GTP exchange protein; Rab3 GEP; RAB3GEP
Rabbit
Protein A
RUO
228355, 8567
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.