missing translation for 'onlineSavingsMsg'
Learn More

Maduramicin ammonium 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30312834
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30312834 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30312834 Supplier Dr. Ehrenstorfer Supplier No. DREA14670000AL100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Maduramicin ammonium salt
CAS 84878-61-5
Analyte or Component Names Maduramicin ammonium salt
Shipping Condition Room Temperature
Molecular Formula C47 H80 O17 . H3 N
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
InChI Formula InChI=1S/C47H80O17.H3N/c1-23-18-24(2)45(9,51)61-36(23)31-19-32(58-35-20-30(53-10)40(55-12)28(6)57-35)42(59-31)44(8)15-14-33(60-44)43(7)16-17-46(64-43)21-29(48)25(3)37(62-46)26(4)38-41(56-13)39(54-11)27(5)47(52,63-38)22-34(49)50;/h23-33,35-42,48,51-52H,14-22H2,1-13H3,(H,49,50);1H3/t23-,24+,25+,26+,27-,28-,29-,30-,31+,32-,33+,35+,36-,37-,38-,39-,40-,41-,42+,43-,44-,45-,46+,47+;/m0./s1
SMILES N.CO[C@H]1C[C@@H](O[C@H]2C[C@@H](O[C@H]2[C@]3(C)CC[C@@H](O3)[C@]4(C)CC[C@]5(C[C@H](O)[C@@H](C)[C@H](O5)[C@@H](C)[C@@H]6O[C@](O)(CC(=O)O)[C@@H](C)[C@H](OC)[C@H]6OC)O4)[C@H]7O[C@](C)(O)[C@H](C)C[C@@H]7C)O[C@@H](C)[C@@H]1OC
IUPAC Name azane;2-[(2R,3S,4S,5R,6S)-6-[(1R)-1-[(2S,5R,7S,8R,9S)-2-[(2R,5S)-5-[(2R,3S,5R)-3-[(2R,4S,5S,6S)-4,5-dimethoxy-6-methyloxan-2-yl]oxy-5-[(2S,3S,5R,6S)-6-hydroxy-3,5,6-trimethyloxan-2-yl]oxolan-2-yl]-5-methyloxolan-2-yl]-7-hydroxy-2,8-dimethyl-1,10-dioxaspiro[4.5]decan-9-yl]ethyl]-2-hydroxy-4,5-dimethoxy-3-methyloxan-2-yl]acetic acid
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 934.16
Quantity 1 mL
Formula Weight 933.5661 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.