missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAF, Mouse anti-Human, Clone: 6B6, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00004094-M01.50ug
This item is not returnable.
View return policy
Description
Sequence: SCRFKRVQQRHVLESEKNQLLQQVDHLKQEISRLVRERDAYKEKYEKLVSSGFRENGSSSDNPSSPEFFITEPTRKLEPSVGYATFWKPQHRVLTSVFTKSpecifications
| MAF | |
| Monoclonal | |
| Unconjugated | |
| v-maf musculoaponeurotic fibrosarcoma oncogene homolog (avian) | |
| MGC71685/c-MAF | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Western Blot | |
| 6B6 | |
| In 1x PBS, pH 7.4 | |
| NM_005360 | |
| MAF | |
| MAF (NP_005351, 304 a.a. ∼ 403 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 4094 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction