missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAFA Recombinant Rabbit Monoclonal Antibody (BLR067G)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA544325
This item is not returnable.
View return policy
Description
Epitope retrieval with citrate buffer pH6.0 is recommended for FFPE cell sections.
MAFA is a transcription factor that binds RIPE3b, a conserved enhancer element that regulates pancreatic beta cell-specific expression of the insulin gene. MAFA cooperates synergistically with NEUROD1 and PDX1. Phosphorylation by GSK3 increases its transcriptional activity and is required for its ocogenic activity. Serves as an ocogene or tumor supressor depending on the cell context.
Specifications
| MAFA | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| BBS with 0.09% sodium azide; pH 8.2 | |
| Q8NHW3 | |
| Mafa | |
| Peptide representing a region between residue 125 and 175 of human MAF bZIP transcription factor A. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| BLR067G | |
| Unconjugated | |
| Mafa | |
| hMafA; MAF bZIP transcription factor A; MAFA; Mafa homolog; pancreatic beta-cell-specific transcriptional activator; RGD1562627; RIPE3b1; RIPE3b1 factor; Transcription factor MafA; transcription factor RIPE3b1; v-maf avian musculoaponeurotic fibrosarcoma oncogene A; v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog A; v-maf musculoaponeurotic fibrosarcoma oncogene family, protein A; v-maf musculoaponeurotic fibrosarcoma oncogene family, protein A (avian); v-maf musculoaponeurotic fibrosarcoma oncogene homolog A | |
| Rabbit | |
| purified | |
| RUO | |
| 389692 | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction