missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAG Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA549646
This item is not returnable.
View return policy
Description
MAG Polyclonal Antibody for Western Blot, Flow
The protein encoded by this gene is a type I membrane protein and member of the immunoglobulin superfamily. It is thought to be involved in the process of myelination. It is a lectin that binds to sialylated glycoconjugates and mediates certain myelin-neuron cell-cell interactions. Two alternatively spliced transcripts encoding different isoforms have been described for this gene.
Specifications
| MAG | |
| Polyclonal | |
| Unconjugated | |
| MAG | |
| 1B236; Brain neuron cytoplasmic protein 3; Gma; MAG; myelin associated glycoprotein; myelin-associated glycoprotein; sialic acid binding Ig-like lectin 4A; sialic acid-binding Ig-like lectin 4a; sialic acid-binding immunoglobulin-like lectin 4A; SIGLEC4A; siglec-4a; S-MAG; S-MAG (C-term.); SPG75 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 17136, 29409, 4099 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P07722, P20916, P20917 | |
| MAG | |
| KLH conjugated synthetic peptide between 439-466 amino acids from the Central region of human MAG. | |
| 400 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction