missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAG Recombinant Rabbit Monoclonal Antibody (8Q7K5)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA542933
This item is not returnable.
View return policy
Description
Positive test controls include: SH-SY5Y, Mouse brain, Rat brain. Immunogen sequence: MIFLTALPLF WIMISASRGG HWGAWMPSSI SAFEGTCVSI PCRFDFPDEL RPAVVHGVWY FNSPYPKNYP PVVFKSRTQV VHESFQGRSR LLGDLGLRNC.
The protein encoded by this gene is a type I membrane protein and member of the immunoglobulin superfamily. It is thought to be involved in the process of myelination. It is a lectin that binds to sialylated glycoconjugates and mediates certain myelin-neuron cell-cell interactions. Two alternatively spliced transcripts encoding different isoforms have been described for this gene.
Specifications
| MAG | |
| Recombinant Monoclonal | |
| 0.53 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P07722, P20916, P20917 | |
| MAG | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human MAG (P20916). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 8Q7K5 | |
| Unconjugated | |
| MAG | |
| 1B236; Brain neuron cytoplasmic protein 3; Gma; MAG; myelin associated glycoprotein; myelin-associated glycoprotein; sialic acid binding Ig-like lectin 4A; sialic acid-binding Ig-like lectin 4a; sialic acid-binding immunoglobulin-like lectin 4A; SIGLEC4A; siglec-4a; S-MAG; S-MAG (C-term.); SPG75 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 17136, 29409, 4099 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction