Learn More
Invitrogen™ MAG Recombinant Rabbit Monoclonal Antibody (PSH02-42)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552084
Description
Tissue Specificity: central and peripheral nervous system Positive Control: Mouse brain (no heat) tissue lysate, mouse cerebellum tissue lysate, rat brain (no heat) tissue lysate, rat cerebellum tissue lysate, human cerebellum tissue, mouse brain tissue, mouse cerebellum tissue, mouse hippocampus tissue, rat brain tissue, rat cerebellum tissue, rat hippocampus tissue. Subcellular Location: Cell membrane. Predicted band size: 69 kDa.
The protein encoded by this gene is a type I membrane protein and member of the immunoglobulin superfamily. It is thought to be involved in the process of myelination. It is a lectin that binds to sialylated glycoconjugates and mediates certain myelin-neuron cell-cell interactions. Two alternatively spliced transcripts encoding different isoforms have been described for this gene.
Specifications
| MAG | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P07722, P20916, P20917 | |
| MAG | |
| Recombinant protein within human mag aa 1- 536. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| PSH02-42 | |
| Unconjugated | |
| MAG | |
| 1B236; Brain neuron cytoplasmic protein 3; Gma; MAG; myelin associated glycoprotein; myelin-associated glycoprotein; sialic acid binding Ig-like lectin 4A; sialic acid-binding Ig-like lectin 4a; sialic acid-binding immunoglobulin-like lectin 4A; SIGLEC4A; siglec-4a; S-MAG; S-MAG (C-term.); SPG75 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 17136, 29409, 4099 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.