missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ MAG/Siglec-4a Antibody (refanezumab) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28053-100ug
This item is not returnable.
View return policy
Description
MAG/Siglec-4a Monoclonal antibody specifically detects MAG/Siglec-4a in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| MAG/Siglec-4a | |
| Monoclonal | |
| Unconjugated | |
| P20916 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| refanezumab | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| GMAsialic acid binding Ig-like lectin 4A, myelin associated glycoprotein, myelin-associated glycoprotein, sialic acid-binding immunoglobulin-like lectin 4A, SIGLEC4A, SIGLEC-4A, S-MAG | |
| Siglec-4a/ MAG | |
| 100 μg | |
| Cell Biology, Neuroscience | |
| 4099 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction