missing translation for 'onlineSavingsMsg'
Learn More

MP BIOMEDICALS MagBeads FastDNA/RNA Kit for Virus Ready-to-Use for MPure-32 96 Preps

Product Code. 30067481 Shop All MP Biomedicals Products
This item is not returnable. View return policy

Product Code. 30067481

Brand: MP BIOMEDICALS 117033300

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MagBeads FastDNA/RNA Kit for Virus is intended for extracting total viral nucleic acid from cell-free/low-cell content biological samples such as body fluids, serum, plasma, soaking solution, tissue homogenate, and culture supernatant using the MPure- 32™ aNAP System.

MagBeads FastDNA/RNA Kit for Virus is based on the purification method of high binding magnetic particles. The sample is lysed and digested with Protease. DNA/RNA is released into the lysate.

After addition of magnetic particles and binding solution:

  • DNA/RNA will be adsorbed on the surface of magnetic particles
  • Impurities such as proteins will be removed without adsorption.

The adsorbed particles are washed with:

  • Washing solution to remove proteins and impurities
  • Ethanol to remove salts

And finally DNA/RNA is eluted with Nuclease Free Water. The purified DNA/RNA is used for RT-PCR and PCR assay.

TRUSTED_SUSTAINABILITY

Specifications

Format Kit
Isolation Technology Magnetic Bead
For Use With (Application) DNA/RNA Extraction
Product Type DNA/RNA Kit for Virus
compliance-icons compliance-icons compliance-icons
Product Identifier
  • (MPure-32) MagBeads FastDNA/RNA Kit for Virus
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P370+P378-In case of fire: Use CO2, powder or waterspray to extinguish.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Proteinase K, Guanidine Hydrochloride, Ethanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.