missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGE 1 Antibody (MA454), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-33094V
This item is not returnable.
View return policy
Description
MAGE 1 Monoclonal specifically detects MAGE 1 in Human, Rat, Canine samples. It is validated for ELISA.
Specifications
| MAGE 1 | |
| Monoclonal | |
| DyLight 405 | |
| Antigen MZ2-E, Cancer/testis antigen 1.1, cancer/testis antigen family 1, member 1, CT1.1melanoma antigen family A 1, MAGE-1 antigen, MAGE1A, MAGE1melanoma antigen MAGE-1, melanoma antigen family A, 1 (directs expression of antigen MZ2-E), melanoma-associated antigen 1, melanoma-associated antigen MZ2-E, MGC9326 | |
| Mouse | |
| 44 kDa | |
| 0.1 mL | |
| Apoptosis, Cancer, Melanoma Cell Markers, Tumor Suppressors | |
| 4100 | |
| Human, Rat, Canine | |
| Purified |
| ELISA | |
| MA454 | |
| ELISA | |
| MAGEA1 | |
| Recombinant full-length human MAGE 1 protein (Uniprot: P43355) | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Recognizes a protein of 42-46kDa, identified as MAGE-1. This monoclonal antibody does not cross-react with MAGE-2, -3, -4, -6 -9, -10, -or -12 protein. Human malignant neoplasms carry rejection antigens that are recognized by the patients' autologous, tumor directed and specific, cytolytic, CD8+ T lymphocyte clones (CTL). The MAGE family of genes codes an important group of antigens. It was identified that melanomas and primary glial brain tumors express common melanoma associated antigens (MAAs). Because MAGE-1 is expressed on a significant proportion of human neoplasms of various histological types (melanoma, brain tumors of glial origin, neuroblastoma, non-small cell lung cancer, breast, gastric, colorectal, ovarian, renal cell carcinomas) and not on normal tissues, the encoded antigen may serve as a marker of early detection and target for cancer immunotherapy. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction