missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGE 1 Antibody (SR1797), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 349.00 - € 449.00
Specifications
| Antigen | MAGE 1 |
|---|---|
| Clone | SR1797 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Flow Cytometry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18667764
|
Novus Biologicals
NBP3-22211-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18629094
|
Novus Biologicals
NBP3-22211-50ul |
50 μg |
€ 349.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MAGE 1 Monoclonal antibody specifically detects MAGE 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ImmunofluorescenceSpecifications
| MAGE 1 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Antigen MZ2-E, Cancer/testis antigen 1.1, cancer/testis antigen family 1, member 1, CT1.1melanoma antigen family A 1, MAGE-1 antigen, MAGE1A, MAGE1melanoma antigen MAGE-1, melanoma antigen family A, 1 (directs expression of antigen MZ2-E), melanoma-associated antigen 1, melanoma-associated antigen MZ2-E, MGC9326 | |
| A synthesized peptide derived from human MAGE 1 (Uniprot #: P43355) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1797 | |
| Western Blot, Flow Cytometry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Melanoma Cell Markers, Tumor Suppressors | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 4100 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title