missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGEA1 Antibody (MZ2E/838), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-11463CL1
This item is not returnable.
View return policy
Description
MAGEA1 Monoclonal antibody specifically detects MAGEA1 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunohistochemistry (Paraffin),Immunofluorescence,Immunocytochemistry/Immunofluorescence
Specifications
| MAGEA1 | |
| Monoclonal | |
| CoraFluor 1 | |
| Antigen MZ2-E, Cancer/testis antigen 1.1, cancer/testis antigen family 1, member 1, CT1.1melanoma antigen family A 1, MAGE-1 antigen, MAGE1A, MAGE1melanoma antigen MAGE-1, melanoma antigen family A, 1 (directs expression of antigen MZ2-E), melanoma-associated antigen 1, melanoma-associated antigen MZ2-E, MGC9326 | |
| Recombinant human MAGEA1 protein (Uniprot: P43355) | |
| 0.1 mL | |
| Apoptosis, Cancer, Melanoma Cell Markers, Tumor Suppressors | |
| 4100 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry/Immunofluorescence | |
| MZ2E/838 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction