missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGEA3 Antibody (21B4), FITC , Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-20166F
This item is not returnable.
View return policy
Description
MAGEA3 Monoclonal antibody specifically detects MAGEA3 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry
Specifications
| MAGEA3 | |
| Monoclonal | |
| FITC | |
| Antigen MZ2-D, Cancer/testis antigen 1.3, CT1.3 MAGE-3 antigen, HIP8, MAGE3 MAGEA6, melanoma antigen family A, 3, melanoma-associated antigen 3, member 3, MGC14613 | |
| The original antibody was isolated from a patient suffering from a tumor expressing a tumor specific antigen like MAGE-A3. | |
| 0.1 mL | |
| Cancer | |
| 4102 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, ELISA, Immunohistochemistry | |
| 21B4 | |
| PBS | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction