missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGEA3 Antibody (OTI1G9), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72570JF549
This item is not returnable.
View return policy
Description
MAGEA3 Monoclonal specifically detects MAGEA3 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| MAGEA3 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MAGEA3 | |
| Full length recombinant protein of human MAGEA3 (NP_005353) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2a |
| Western Blot, Immunohistochemistry | |
| OTI1G9 | |
| Western Blot, Immunohistochemistry | |
| Antigen MZ2-D, Cancer/testis antigen 1.3, CT1.3 MAGE-3 antigen, HIP8, MAGE3 MAGEA6, melanoma antigen family A, 3, melanoma-associated antigen 3, member 3, MGC14613 | |
| Mouse | |
| Affinity Purified | |
| Cancer | |
| 4102.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction