missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGEA3 Monoclonal Antibody (OTI2F10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800802
This item is not returnable.
View return policy
Description
This gene is a member of the MAGEA gene family. The members of this family encode proteins with 50 to 80% sequence identity to each other. The promoters and first exons of the MAGEA genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEA genes are clustered at chromosomal location Xq28. They have been implicated in some hereditary disorders, such as dyskeratosis congenita.Specifications
| MAGEA3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P43357 | |
| MAGEA3 | |
| Full length human recombit protein of human MAGEA3 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2F10 | |
| Unconjugated | |
| MAGEA3 | |
| CT1.3, HIP8, HYPD, MAGE3, MAGEA6 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 4102 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction