missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGEA3 Monoclonal Antibody (OTI2F10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800802S
This item is not returnable.
View return policy
Description
MAGEA3 Monoclonal antibody specifically detects MAGEA3 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| MAGEA3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P46013 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4288 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2F10 | |
| Unconjugated | |
| Mki67 | |
| Antigen MZ2-D; cancer/testis antigen 1.3; cancer/testis antigen family 1, member 3; CT1.3; HIP8; HYPD; MAGE family member A3; MAGE3; MAGE-3 antigen; MAGEA3; Mage-a3; MAGEA6; melanoma antigen family A, 3; melanoma antigen family A3; melanoma antigen, family A, 3; melanoma-associated antigen 3; MGC14613 | |
| Human recombinant protein fragment corresponding to amino acids 2924-3256 of human MKI67 produced in E.coli | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction