missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGEA4 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41318CL1
This item is not returnable.
View return policy
Description
MAGEA4 Polyclonal antibody specifically detects MAGEA4 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| MAGEA4 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| MAGE-X2 antigen, melanoma antigen family A, 4, melanoma-associated antigen 4, member 4 | |
| MAGEA4 antibody was raised against a 16 amino acid peptide near the amino terminus of human MAGEA4. The immunogen is located within the first 50 amino acids of MAGEA4. Amino Acid Squence: VGAQAPTTEEQEAAVS | |
| 0.1 mL | |
| Cancer | |
| 4103 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction