missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGEA4 Antibody (CPTC-MAGEA4-1), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79861APC
This item is not returnable.
View return policy
Description
MAGEA4 Monoclonal antibody specifically detects MAGEA4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array
Specifications
| MAGEA4 | |
| Monoclonal | |
| APC | |
| MAGE-X2 antigen, melanoma antigen family A, 4, melanoma-associated antigen 4, member 4 | |
| Recombinant full-length human MAGEA4 protein (Uniprot: P43358) | |
| 0.1 mL | |
| Cancer | |
| 4103 | |
| Store at 4°C in the dark. | |
| IgG2c κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array | |
| CPTC-MAGEA4-1 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction