missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGEA4 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41318H
This item is not returnable.
View return policy
Description
MAGEA4 Polyclonal antibody specifically detects MAGEA4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| MAGEA4 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| HRP | |
| MAGE-X2 antigen, melanoma antigen family A, 4, melanoma-associated antigen 4, member 4 | |
| MAGEA4 antibody was raised against a 16 amino acid peptide near the amino terminus of human MAGEA4. The immunogen is located within the first 50 amino acids of MAGEA4. Amino Acid Squence: VGAQAPTTEEQEAAVS | |
| 0.1 mL | |
| Cancer | |
| 4103 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction