missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAGEA5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA542849
This item is not returnable.
View return policy
Description
Peptide sequence: MSLEQKSQHC KPEEGLDTQE EALGLVGVQA ATTEEQEAVS SSSPLVPGTL Sequence homology: Human: 100%.
This gene is a member of the MAGEA gene family. The members of this family encode proteins with 50 to 80% sequence identity to each other. The promoters and first exons of the MAGEA genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEA genes are clustered at chromosomal location Xq28. They have been implicated in some hereditary disorders, such as dyskeratosis congenita. This MAGEA gene encodes a protein that is C-terminally truncated compared to other family members, and this gene can be alternatively interpreted to be a pseudogene. The protein is represented in this Gene record in accordance with the assumed protein-coding status defined in the literature. Read-through transcription exists between this gene and the upstream melanoma antigen family A, 10 (MAGEA10) gene. [provided by RefSeq, Oct 2011].
Specifications
| MAGEA5 | |
| Polyclonal | |
| Unconjugated | |
| MAGEA5 | |
| Cancer/testis antigen 1.5; cancer/testis antigen family 1, member 5; CT1.5; MAGE family member A5; MAGE5; MAGE-5 antigen; MAGE-5a antigen; MAGE-5b antigen; MAGEA4; Magea5; Mage-a5; melanoma antigen family A, 5; melanoma antigen family A5; melanoma antigen, family A, 5; melanoma-associated antigen 5; MGC129526 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 4104 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P43359 | |
| MAGEA5 | |
| Synthetic peptide directed towards the N-terminal of human MAGEA5 (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction