missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGEA9 Antibody (OTI1D8), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72572AF750
This item is not returnable.
View return policy
Description
MAGEA9 Monoclonal specifically detects MAGEA9 in Human samples. It is validated for Western Blot.
Specifications
| MAGEA9 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| MAGEA9 | |
| Full length recombinant protein of human MAGEA9 (NP_005356) produced in HEK293T cells. | |
| 0.1 mL | |
| 4108.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI1D8 | |
| Western Blot | |
| Cancer/testis antigen 1.9, CT1.9MAGEA9B, MAGE-9 antigen, MAGE9cancer/testis antigen family 1, member 9, MAGEA9A, melanoma antigen family A, 9, melanoma-associated antigen 9, MGC8421 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction